Restek™ CarboPrep Cartridges
Our manufacturing process minimizes variability and improves recovery and cleanup procedures.
Manufacturer: Restek™ 26087
 View more versions of this product
Catalog No. 06-713-148
  Check Availability
Add to Cart
    Specifications
    Product Type SPE cartridge
    Bed Weight (Metric) 500mg
    Cartridge Material Silica
    For Use With (Application) Sample extraction of organic compounds and extract cleanup of matrix interferences
    Packing Material CarboPrep 200
    Surface Area (Metric) 200m2/g
    Tube Size 6mL, 500mg
    Volume (Metric) 6mL
  • Description & Specifications
Related Products
Product Content Feedback