Restek™ CarboPrep Cartridges
Our manufacturing process minimizes variability and improves recovery and cleanup procedures.
Manufacturer: Restek™ 26088
 View more versions of this product
Catalog No. 06-713-149
  Check Availability
Add to Cart
    Specifications
    Product Type SPE cartridge
    Tube Size 3mL, 250mg
    Bed Weight (Metric) 250mg
    Cartridge Material Silica
    For Use With (Application) Sample extraction of organic compounds and extract cleanup of matrix interferences
    Packing Material CarboPrep 200
    Surface Area (Metric) 200m2/g
    Volume (Metric) 3mL
  • Description & Specifications
Related Products
Product Content Feedback