Restek™ Chiral Column Kits Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Restek™ 13190
 View all options for this product
Catalog No. 06-710-013
  Check Availability
Add to Cart
    For Use With Enantiomeric profile and conformational identification of individual chiral compounds
    Length (Metric) 30m
    Technique GC
    Diameter (Metric) Inner 0.25mm
    Includes Rt™-betaDEXcst column and Rt™-betaDEXsm column
  • Description & Specifications
Related Products
Product Content Feedback