Restek™ Chiral Column Kits Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Restek™ 13196
 View more versions of this product
Catalog No. 06-710-019
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With Enantiomeric profile and conformational identification of individual chiral compounds
    Technique GC
    Diameter (Metric) Inner 0.25mm
    Includes Rt™-betaDEXsm column, Rt™-betaDEXse column, Rt™-betaDEXsa column and Rt™-betaDEXsp column
    Length (Metric) 30m
  • Description & Specifications
Related Products
Product Content Feedback