Restek™ Chromosorb Packings
Manufacturer: Restek™ 25639
 View all options for this product
Catalog No. 06-713-027
  Check Availability
Add to Cart
    Material Chromosorb PAW/DMDCS
    Particle Size 80/100 Mesh
    Quantity 100g
    Product Type Packing Material
  • Description & Specifications
Related Products
Product Content Feedback