Restek™ Chromosorb Packings
Manufacturer: Restek™ 25651
 View all options for this product
Catalog No. 06-713-039
  Check Availability
Add to Cart
    Material Chromosorb GAW/DMDCS
    Particle Size 45/60 Mesh
    Quantity 100g
    Product Type Packing Material
  • Description & Specifications
Related Products
Product Content Feedback