Learn More
Restek™ Chromosorb Packings
Packed column packing material; packing only. Chromosorb diatomaceous earth supports are specially sieved to remove fines and ensure tight particle distribution.
Catalog Number | Mfr. No. | Particle Size | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Particle Size | Material | Price | Quantity & Availability | ||||
06-713-019
|
Restek™
25631 |
80/100 Mesh | Chromosorb PNAW | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
06-713-020
|
Restek™
25632 |
100/120 Mesh | Chromosorb PNAW | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
06-713-022
|
Restek™
25634 |
60/80 Mesh | Chromosorb PAW | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
06-713-023
|
Restek™
25635 |
80/100 Mesh | Chromosorb PAW | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
06-713-041
|
Restek™
25653 |
80/100 Mesh | Chromosorb GAW/DMDCS | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
06-713-053
|
Restek™
25665 |
60/80 Mesh | Chromosorb WAW/DMDCS | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Chromosorb P support can hold up to 30 weight% of liquid stationary phase, making it the highest loading support available
- Chromosorb G support is the hardest support available and has the lowest surface area of all the diatomaceous earth supports
- Chromosorb W support is a flux-calcinated diatomite. This solid support is very fragile but offers the highest inertness of all diatomaceous earth supports.
Specifications
Packing Material |