Restek™ Chromosorb Packings
Manufacturer: Restek™ 25608
 View all options for this product
Catalog No. 06-712-996
  Check Availability
Add to Cart
    Material Chromosorb 101
    Max. Temperature 325°C
    Particle Size 60/80 Mesh
    Quantity 50g
    Product Type Packing Material
  • Description & Specifications
Related Products
Product Content Feedback