Restek™ Column End-Fittings
These 0.25 in. compression end-fittings are compatible with 0.0625 in. connecting tubing using Valco and Parker fittings.
Manufacturer: Restek™ 25078
 View more versions of this product
Catalog No. 06-712-649
  Check Availability
Add to Cart
Related Products
Product Content Feedback