Learn More
Restek™ Critical Purity Switchover System for Noncorrosive Service
Allow the replacement of a depleted gas source without interrupting the gas supply to the instrument or process
Manufacturer:  Restek™ 21593580
 View more versions of this product
Catalog No. 06-711-679
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Provide a continuous supply of high purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time
Line regulator, Stainless Steel pigtails with armor casing
Stainless Steel
Gas regulator
Argon, Helium, Nitrogen
580
200/170psi