Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Cyclo Vu-Tight DI Liner (1/4-inch OD)

Visually observe the Press-Tight connection between the column end and liner.

Manufacturer:  Restek™ 20787

 View more versions of this product

Catalog No. 06-711-041


  Check Availability
Add to cart

Specifications

Description

  • Fits 0.32 and 0.53mm ID capillary columns (column ODs from 0.5 to 0.8mm)
  • Slotted top prevents obstruction of carrier gas flow
  • Operate in the direct injection mode
  • Spiral bore prevents nonvolatile residue from contaminating the column
  • Ideal for dirty samples
  • O.D.: 0.25 in.
Description & Specifications

Specifications

Cyclo Vu-tight direct injection liner
Each
0.63cm
0.32 and 0.53mm I.D. Capillary Columns
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit