Learn More
Restek™ Detector Lamps
Used for waters detectors or for Agilent HPLC Systems
Manufacturer:  Restek™ 25404
 View more versions of this product
Catalog No. 06-712-914
Add to cart
Description
Description
  • Standard lamps have nominal 1,000-hour life
Specifications
Specifications
Xenon Lamp w/ Holder or Mirror
470 Water Detector
1 Chromatography Detector Lamp
Safety and Handling
Safety and Handling
ShelfLife : 1000 hr.