Restek™ Detector Lamps

Used for waters detectors or for Agilent HPLC Systems

Manufacturer: Restek™ 25403

 View all options for this product

Catalog No. 06-712-913

  Check Availability
Add to Cart


    Product Type Deuterium lamp
    For Use With (Equipment) 480, 481 Water detector
    Includes 1 Chromatography Detector Lamp
    Shelf Life 1000 hr.

    • Standard lamps have nominal 1,000-hour life

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit