Restek™ Detector Lamps

Used for waters detectors or for Agilent HPLC Systems

Manufacturer: Restek™ 25405

 View more versions of this product

Catalog No. 06-712-915

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) 474 Water detector
    Includes 1 Chromatography Detector Lamp
    Shelf Life 1000 hr.
    Product Type Xenon lamp

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit