Restek™ DI Liners for Shimadzu GCs

For Shimadzu 17A, 2010, and 2014 GCs equipped with split/splitless inlets.

Manufacturer: Restek™ 21713

 View more versions of this product

Catalog No. 06-718-597

  Check Availability
Add to Cart

    Specifications

    Product Type IP Uniliner™ inlet liner
    For Use With (Equipment) Shimadzu™ 17A, 2010, and 2014 GCs equipped with split/splitless inlets
    Packing Material Deactivated Wool
    Diameter (Metric) Inner 0.35cm
    Diameter (Metric) Outer 0.5cm
    Length (Metric) 9.5cm
    Liner Material Borosilicate Glass
    Quantity Each

    • Trace, dirty, high MW, active samples, high recovery and linearity
    • Geometry: Uniliner
    • Material: Borosilicate Glass
    • Deactivation: Intermediate Polarity (IP)
    • Packing: Deactivated Wool
    • I.D.: 3.5mm
    • OD: 5mm
    • Length: 95mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit