Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ DI Liners for Shimadzu GCs

For Shimadzu 17A, 2010, and 2014 GCs equipped with split/splitless inlets.

$258.00

Specifications

Diameter (Metric) Inner 0.35 cm
Diameter (Metric) Outer 0.5 cm
Liner Material Borosilicate Glass
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Length (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Length (Metric) Quantity Price Quantity & Availability  
06-718-611
View Documents
Restek™
21719
Shimadzu 17 A, 2010, and 2014 GCs Equipped with Split/Splitless Inlets 9.5 cm 5/Pk.
Pack of 5 for $258.00
Add to cart
 
06-718-597
View Documents
Restek™
21713
Shimadzu 17 A, 2010, and 2014 GCs Equipped with Split/Splitless Inlets 9.5 cm Each This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Trace, dirty, high MW, active samples, high recovery and linearity
  • Geometry: Uniliner
  • Material: Borosilicate Glass
  • Deactivation: Intermediate Polarity (IP)
  • Packing: Deactivated Wool
  • I.D.: 3.5mm
  • OD: 5mm
  • Length: 95mm
Specifications

Specifications

0.35 cm
0.5 cm
Borosilicate Glass
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit