Learn More
Restek™ Direct Injection (DI) Uniliner Liners for Agilent 5890 Packed Column GC
Use with 0.25, 0.32, and 0.53mm ID capillary columns
Manufacturer: Restek™ 20965214.5
Description
- Press-Tight™ taper forms dead volume free connection to column
- Minimizes solvent and peak tailing
- Can be removed from the septum nut weldment
Specifications
1.7 mm | |
Agilent 5890 Packed Column GC, 0.25, 0.32, and 0.53 mm I.D. Capillary Columns | |
5/Pk. |
0.3 cm | |
9.3 cm |