Learn More
Restek™ Double Gooseneck Splitless Liners for Thermo™ TRACE™ and Focus™ SSL
Double gooseneck design for increased performance with larger sample introductions.
Manufacturer:  Restek™ 20953214.5
 View more versions of this product
Catalog No. 06-718-244
  Check Availability
Add to cart
Specifications
Description
4.0 mm ID Double Taper Inlet Liner for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets
Description & Specifications
Specifications
Siltek
8.0mm
Pk. of 5
Siltek Deactivated
105mm