Restek™ Double Gooseneck Splitless Liners for Thermo™ TRACE™ and Focus™ SSL
Double gooseneck design for increased performance with larger sample introductions
Manufacturer: Restek™ 20953210.5
 View more versions of this product
Catalog No. 06-711-281
  Check Availability
Add to Cart
    Specifications
    Deactivation Base Deactivated
    Quantity 5/Pk.
    Diameter (Metric) Outer 8.0mm
    Length (Metric) 105mm
    Closure Liner Glass
  • Description & Specifications
Related Products
Product Content Feedback