Learn More
Restek™ Dual-Column mini-Lam Direct Injection Tee
For 0.25 in. packed column inlets.
Manufacturer:  Restek™ 20435
 View more versions of this product
Catalog No. 06-710-635
Add to cart
Description
Description
  • Inverted cup design for complete sample vaporization
  • Permits larger sample volumes
  • Incorporates a Press-Tight taper in each outlet leg, to make a perfect, dead-volume-free connection to each analytical column (ODs ranging from 0.4 to 0.8mm)
  • Allows visual confirmation of the column connection
  • Open-top design makes it easy to pack with glass wool, keeping sample residue from contaminating the cup
  • Includes all fittings and ferrules
Specifications
Specifications
Replacement Tee for Dual Column mini-Lam Direct Injection Tee Kit
Replacement Part for 0.25 in. Packed Column Inlets