Learn More
Restek™ Dual-Column mini-Lam Direct Injection Tee
SDP
For 0.25 in. packed column inlets.
$135.00 - $234.00
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
06-710-637
SDP
View Documents
Restek™
20436
Dual Column mini-Lam Direct Injection Tee Kit for 1/4-inch packed column inlets
Each for $234.00
Add to cart
06-710-635
SDP
View Documents
Restek™
20435
Replacement Tee for Dual Column mini-Lam Direct Injection Tee Kit
Each for $135.00
Add to cart
Description
Description
  • Inverted cup design for complete sample vaporization
  • Permits larger sample volumes
  • Incorporates a Press-Tight taper in each outlet leg, to make a perfect, dead-volume-free connection to each analytical column (ODs ranging from 0.4 to 0.8mm)
  • Allows visual confirmation of the column connection
  • Open-top design makes it easy to pack with glass wool, keeping sample residue from contaminating the cup
  • Includes all fittings and ferrules