Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ ELCD Supplies
SDP

Catalog No. p-64328 Shop All Restek Corporation Products
Click to view available options
Diameter (English) Inner:
0.781 in.
1.375 in.
Model:
103 C
108

UV energy emitting PID lamps for use with ELCD detectors.

Model 108

  • Offers both 10.0 and 10.6 eV potential
  • Base Dia.: 0.781 in.
  • Used in Tracor, OI, and Baseline instruments

Model 103C

  • Base Dia.: 1.375 in.
  • Used in HNU and SRI detectors

Specifications

Type Lamp
Diameter (English) Inner 1.375 in.
Model 103 C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.