Learn More
Restek™ Empty Chromatography Columns
HPLC column assembly made from 316 stainless steel tubing is complete with end-fittings, frits, nuts, and ferrules.
Manufacturer:  Restek™ 25129
 View more versions of this product
Catalog No. 06-712-696
  Check Availability
Add to cart
Specifications
Description
  • Preassembled prior to shipment, unless otherwise requested
  • Internal 0.25 in. seats are compatible with Valco and Parker fittings
  • O.D.: 0.25 in.
Description & Specifications
Specifications
Empty chromatography column Valco™ and Parker™ fitting
250mm
3.2mm
End fittings, frits, nuts and ferrules
Column Assembly
316 Stainless Steel
0.25mm