Learn More
Restek™ FAMEWAX Capillary Columns
Application-specific columns for FAMEs
Supplier: Restek™ 124996850
Description
- Crossbond™ polyethylene glycol
- Meet AOCS and AOAC method requirements
- Elution order of complex polyunsaturated fatty acid methyl esters (FAMEs) is comparable to that on other Carbowax™ columns, but baseline resolution is achieved in significantly less time
Specifications
0.53 mm | |
20°C to 250°C | |
30 m |