Learn More
Restek™ Filter and Baseplate Kits
SDP
Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
Supplier:  Restek™ 22021
 View more versions of this product
Catalog No. 06-711-825
Add to Cart
Description
Description
Carrier Gas Cleaning Kit
  • Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
  • Triple filter for oxygen, moisture, and hydrocarbon impurities (06-711-824)
Fuel Gas Purification Kit
  • Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
  • Fuel gas filter for moisture and hydrocarbon impurities (06-711-826)
Specifications
Specifications
Fuel Gas Purification Kit
Filtering for moisture and hydrocarbon impurities
Mounting base plate, 0.12 in. inlet/outlet fittings, and hydrocarbon/moisture filter
Filter and Baseplate Kit
Product Suggestions
Product Suggestions