Restek™ Filter and Baseplate Kits
Single-position mounting baseplate with 0.125 in. inlet/outlet fittings (06-711-828)
Manufacturer: Restek™ 22019
 View more versions of this product
Catalog No. 06-711-823
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Filtering oxygen, moisture, and hydrocarbon impurities
    Includes Mounting base plate, 0.12 in. inlet/outlet fittings, and oxygen/moisture/hydrocarbon triple filter
    Style Filter and Baseplate Kit
    Product Type Carrier gas cleaning kit
  • Description & Specifications
Related Products
Product Content Feedback