Learn More
Restek™ Fittings, Parker (Treated): End Reducer Tubes
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
Manufacturer:  Restek™ 22882
 View more versions of this product
Catalog No. 06-712-087
  Check Availability
Add to cart
Specifications
Description
  • Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Description & Specifications
Specifications
Tube End Reducer
Silcosteel-CR Coated
1/8 in. to 1/4 in.