Restek™ Fittings, Parker (Treated): NPT Female Connectors
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
Manufacturer: Restek™ 21559
 View more versions of this product
Catalog No. 06-711-670
  Check Availability
Add to Cart
    Specifications
    Material Sulfinert Coated
    Connections 1/8 in. to 1/4 in.
    Type NPT Female Connector
    • Material: Siltek/Sulfinert-Treated
  • Description & Specifications
Related Products
Product Content Feedback