Restek™ Fittings, Parker (Treated): Unions
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
Manufacturer: Restek™ 22887
 View more versions of this product
Catalog No. 06-712-092
  Check Availability
Add to Cart
    Specifications
    Material Silcosteel-CR Coated
    Type NPT Male Union
    For Use With Chromatography connectors
    • Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
  • Description & Specifications
Related Products
Product Content Feedback