Learn More
Restek™ Gas-Specific Purifiers
Ideal for removing moisture, hydrocarbons and oxygen from gas chromatography gas supplies
Manufacturer:  UOP Mat Sen P300 1
 View more versions of this product
Catalog No. 05-730-5
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.12 in. Fitting
3.8cm
500mL/min.
Nitrogen
57.15cm
1.5 in.
Gas purifier
Ideal for removing moisture, Hydrocarbons and Oxygen from gas chromatography gas supplies
22.5 in.
1000psi