Learn More
Restek™ Gerstel GRAPHPACK 3D/2 Connectors
SDP
Complete system quickly and reliably makes a leak-tight, low-dead-volume connection.
Supplier:  Restek™ 20273
 View more versions of this product
Catalog No. 06-710-558
Add to cart
Description
Description
  • Central component is a metal-jacketed graphite ferrule
  • Ideal seal for GC applications
Note
Use only with GRAPHPACK 3D/2 ferrules
Specifications
Specifications
0.45 mm to 0.70 mm ID Column
Connector