Learn More
Restek Gerstel GRAPHPACK 3D/2 Connectors
Complete system quickly and reliably makes a leak-tight, low-dead-volume connection.
$257.20 - $262.53
Catalog Number | Mfr. No. | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With | Price | Quantity & Availability | |||||
06-710-557
|
Restek
20272 |
0.25 mm to 0.32 mm ID Column |
Each for $257.20
|
|
06-710-558
|
Restek
20273 |
0.45 mm to 0.70 mm ID Column |
Each for $262.53
|
|
Description
- Central component is a metal-jacketed graphite ferrule
- Ideal seal for GC applications
Note
Use only with GRAPHPACK 3D/2 ferrules
Specifications
Connector |