Learn More
Restek™ Guard Cartridge Holder
Use with any HPLC column by connecting it with a short piece of 0.0625 in. tubing, appropriate nuts and ferrules, or finger-tight fittings.
Manufacturer:  Restek™ 25060
 View more versions of this product
Catalog No. 06-712-632
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
20mm Trident™ guard cartridge
Guard Cartridge Holder