Restek™ Guard Cartridge Holder
Use with any HPLC column by connecting it with a short piece of 0.0625 in. tubing, appropriate nuts and ferrules, or finger-tight fittings.
Manufacturer: Restek™ 25061
 View more versions of this product
Catalog No. 06-712-633
  Check Availability
Add to Cart
    Specifications
    Type Guard Cartridge Holder
    For Use With 200mm Guard cartridge
  • Description & Specifications
Related Products
Product Content Feedback