Learn More
Restek™ Hamilton Autosampler Syringes for PerkinElmer Autosystem GCs
For PerkinElmer Autosystem and Clarus 500 GCs.
$99.00
Specifications
Needle Point Number 3
Volume (Metric) 5 µL
View More Specs
Products 1
Catalog Number Mfr. No. Needle Gauge Price Quantity & Availability  
Catalog Number Mfr. No. Needle Gauge Price Quantity & Availability  
06-712-577
View Documents
Restek™
24952
26 G
Each for $99.00
Add to cart
Description
Description
  • Meets critical autosampler performance needs
  • Unique plunger design eliminates bending and seizing
  • Needle point styles withstands multiple, rapid injections through a septum
  • Volume: 5µL
  • Gauge: 26
  • Length: 2.756 in.
  • Point Style: 3
Specifications
Specifications
3
5 µL