Learn More
Handheld Electronic Leak Detector Accessories from Restek™
Protects data and analytical columns from costly repairs
Supplier: Restek Corporation 22657
Alert:
Electronic leak detector determines trace amounts of gas in a GC environment only. Unit is not designed for determining combustible gas leaks. Avoid using liquid leak detectors in a capillary system: liquids can be drawn into the system.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Includes: Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs)
Description
- Optimized sample flow path
- Sleek ergonomic design
- Handy probe storage
- Long-lasting battery: up to 6 hours of continuous use
- Automatic shut-off capabilities
- Soft-sided carrying case designed for storage in a smaller space
- Small probe detector useful to verify hard-to-reach leaks
Certifications
CE marked
Compliance
WEEE, RoHS
Specifications
Handheld Leak Detector | |
CE, WEEE, RoHS | |
Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs) |
Soft Side Carrying Case | |
Protects data and analytical columns from costly repairs |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.