Restek™ High-Capacity Indicating Oxygen Traps
High capacity design for long life; usable with all carrier gases
Manufacturer: Restek™ 20625
 View more versions of this product
Catalog No. 06-710-857
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Application) All carrier gases
    For Use With (Equipment) High capacity indicating Oxygen trap, with either 1/8 in. or 1/4 in. fitting
    Capacity (English) 0.07 oz.
    Product Type Replacement Cartridge
  • Description & Specifications
Related Products
Product Content Feedback