Restek™ HPLC Stainless Steel Capillary Tubing Assemblies
For Agilent HPLC Systems.
Manufacturer: Restek™ 26530
 View more versions of this product
Catalog No. 06-728-955
  Check Availability
Add to Cart
    Specifications
    Product Type Seat Capillary, SS Tubing
    Diameter (English) Inner 0.006 in.
    Finish Smooth
    For Use With (Application) HPLC systems
    For Use With (Equipment) Agilent HPLC systems
    Length (Metric) N/A
    Material N/A
    Model 1050
    • Precut, micropolished tubing and preseated fittings for quick, easy maintenance of your Agilent HPLC systems
    • Material: Stainless steel
  • Description & Specifications
Related Products
Product Content Feedback