Learn More
Restek™ Inert PEEK™ Tubing
Replaces stainless steel, PTFE, and ETFE tubing
Manufacturer:  Restek™ 25320
 View more versions of this product
Catalog No. 06-712-860
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Pink Dash Stripe
0.625 in.
PEEK fingertight or flangeless fittings
1m
7000psi
482°F
0.0025 in.
0.158cm
3.28 ft.
PEEK
Inert PEEK tubing
250°C
Accessories