Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Inlet Liner Seals for Varian 1177 Injectors

For Agilent and Bruker/Varian 1177 Injectors.

Manufacturer:  Restek™ 20298

 View more versions of this product

Catalog No. 06-717-677


  Check Availability
Add to cart

Specifications

Description

  • High thermal stability at injection port temperatures up to 450°C
  • For split or splitless liners
Description & Specifications

Specifications

O-ring
842°F
0.65cm
Graphite
10/Pack
450°C
Split liners, Agilent and bruker/varian 1177 Injectors
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit