Learn More
Restek™ Inlet Liners for Basic Organic Analysis for Agilent™ GCs: Cyclosplitter
Specifically designed to ensure a highly inert GC pathway for basic compounds
Manufacturer:  Restek™ 20708210.25
 View more versions of this product
Catalog No. 06-717-804
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Cyclosplitter™ base-Deactivated inlet liner
0.4cm
7.85cm
Borosilicate Glass
25/Pack
0.63cm
Agilent GCs equipped with split/splitless inlets