Restek™ Inlet Liners for Basic Organic Analysis for Agilent™ GCs: Split Liner

Specifically designed to ensure a highly inert GC pathway for basic compounds

Manufacturer: Restek™ 20781211.1

 View all options for this product

Catalog No. 06-711-028


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.


    Packing Material Base Deactivated Wool
    Diameter (Metric) Inner 0.4cm
    Diameter (Metric) Outer 0.63cm
    Length (Metric) 7.85cm
    For Use With (Equipment) Agilent GCs equipped with split/splitless inlets
    Liner Material Borosilicate Glass
    Quantity Each
    Product Type Split base-Deactivated straight inlet liner

    Split liner design with base-deactivated wool packing material.

    Base-deactivation process:

    • Bonds basic functional groups to the analytical surface to reduce adsorption and priming effects
    • I.D. 4mm
    • Improves peak shape, response and reproducibility when compared to standard deactivation

    • Base-deactivated wool prevents the vaporized sample from contacting the adsorptive hot metal inlet disc surface

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit