Learn More
Restek™ Inlet Liners for Split Injection
Fully deactivated to prevent degradation or adsorption of active compounds
Manufacturer:  Restek™ 20781213.1
 View more versions of this product
Catalog No. 06-711-029
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Siltek
Inlet liner
Deactivated Wool
6.3mm
Agilent GCs equipped with split/splitless inlets
Straight
Ea.
4.0mm
7.85cm
Borosilicate Glass