Restek™ Inlet Liners for Splitless Injection—Gooseneck Splitless
Useful for splitless mode of operation and slow injections, small (<2µL) sample volumes, and high-boiling point solvent systems
Manufacturer: Restek™ 20799202.5
 View more versions of this product
Catalog No. 06-717-994
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type IP gooseneck splitless Single taper inlet liner; For trace samples greater than 2μL
    Packing Material Glass Wool Prepacked Inlet Liner
    Diameter (Metric) Inner 0.4cm
    Diameter (Metric) Outer 0.65cm
    Length (Metric) 78.5mm
    For Use With (Equipment) Agilent GCs equipped with split/splitless inlets
    Quantity 5/Pk.
  • Description & Specifications
Related Products
Product Content Feedback