Learn More
Restek™ Inlet Liners for Splitless Injection—Siltek™ Gooseneck Splitless
Useful for splitless mode of operation and slow injections, small (<2µL) sample volumes, and high-boiling point solvent systems
Manufacturer:  Restek™ 20800214.25
 View more versions of this product
Catalog No. 06-718-005
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Siltek™ Single gooseneck splitless inlet liner
0.4cm
7.85cm
Borosilicate Glass
25/Pack
0.65cm
Agilent GCs equipped with split/splitless inlets