Restek™ Inlet Seals
Replacement seals for Agilent™ 5890/6890/6850 split/splitless injection ports
Manufacturer: Restek™ 20391
 View more versions of this product
Catalog No. 06-717-747
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Agilent 5890/6890/6850 split/splitless
    For Use With (Application) Inlet Seals
    Material Stainless-Steel
    Diameter (Metric) Inner 0.25/0.32mm
  • Description & Specifications
Related Products
Product Content Feedback