Restek™ Kontes™ Membrane Filtration Glassware
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
Manufacturer: Restek™ KT9538250000
 View more versions of this product
Catalog No. 06-717-524
  Check Availability
Add to Cart
    Specifications
    Description Funnel, Flask
    For Use With (Equipment) Microfiltration Apparatus
    Capacity (Metric) 300mL, 1000mL
    Material Glass
  • Description & Specifications
Related Products
Product Content Feedback