Learn More
Restek™ Liners for Thermo™ TRACE™ and Focus™ SSL: Splitless with CarboFrit
For Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets.
Manufacturer:  Restek™ 20946209.5
 View more versions of this product
Catalog No. 06-718-224
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Splitless Single taper liner
CarboFrit™
0.8cm
Thermo Trace™, 8000 series and focus GCs equipped with SSL inlets
5/Pk.
0.5cm
10.5cm
Borosilicate Glass