Learn More
Restek™ Low-Pressure, CPI Inlet Filter for Mobile Phase Reservoir
A 316 stainless steel knurled cap and Tefzel CPI ferrule seal to 0.125 in. OD PTFE tubing when finger-tightened onto the precision-machined filter holder.
Manufacturer:  Restek™ 25010
 View more versions of this product
Catalog No. 06-719-059
  Check Availability
Add to cart
Specifications
Description
  • Replaceable filter element
  • Standard 10µm porosity protects delicate pump components from particles but introduces very little pressure drop
  • May be used as a helium sparging diffuser
  • O.D.: 0.125 in.
Description & Specifications
Specifications
Replacement Element
Low-Pressure CPI Inlet Filter for Mobile Phase Reservoir
PTFE
10μm