Restek™ Methane Cylinder Kit
Methane kit plus replacement parts
Manufacturer: Restek™ 20198
 View all options for this product
Catalog No. 06-717-609
  Check Availability
Add to Cart
    Product Type Replacement Septa
    For Use With (Application) For optimizing linear velocity
    For Use With (Equipment) For syringe adaptor, Methane cylinder
    For optimizing linear velocity
    Methane Cylinder Kit
    • Set the column flow rate by injecting methane and optimizing linear velocity
    • Uses the peferred method for establishing reproducible retention times (ASTM Method E1510-93)
    • Includes
    • Scotty 14 cylinder containing 1% methane in helium
    • MINICYL regulator
    • Syringe adaptor
    • Package of twenty-five septa for the adaptor
  • Description & Specifications
Related Products
Product Content Feedback